Ethyl-2-isothiocyanatobenzoat 98+%

Lieferant: Apollo Scientific

Synonyme: o-Ethoxycarbonylphenylisothiocyanat

OR315248-1G OR315248-5G
APOSOR315248-1GEA 91.5 EUR
APOSOR315248-1G APOSOR315248-5G
Ethyl-2-isothiocyanatobenzoat 98+%
Ethyl-2-isothiocyanatobenzoat
Formel: C₁₀H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00041063
CAS-Nummer: 99960-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR