Ethyl-2-isothiocyanatobenzoat 98+%
Lieferant: Apollo Scientific
Synonyme:
o-Ethoxycarbonylphenylisothiocyanat
OR315248-1G
OR315248-5G
APOSOR315248-1GEA
91.5
EUR
APOSOR315248-1G
APOSOR315248-5G
Ethyl-2-isothiocyanatobenzoat 98+%
Ethyl-2-isothiocyanatobenzoat
Formel:
C₁₀H₉NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041063 CAS-Nummer: 99960-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...