2,6-Dichlorphenylisothiocyanat

Lieferant: Apollo Scientific

Synonyme: 1,3-Dichloro-2-isothiocyanatobenzene

OR315263-5G
APOSOR315263-5GEA 102 EUR
APOSOR315263-5G
2,6-Dichlorphenylisothiocyanat
2,6-Dichlorphenylisothiocyanat
Formel: C₇H₃Cl₂NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00041056
CAS-Nummer: 6590-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR