(-)-Dimethyl-L-malat 98%

Lieferant: Apollo Scientific

OR315283-100G
APOSOR315283-100GEA 0 EUR
APOSOR315283-100G
(-)-Dimethyl-L-malat 98%
(-)-Dimethyl-L-malat
Formel: C₆H₁₀O₅
Siedepunkt: 104…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066215
CAS-Nummer: 617-55-0
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR