(R)-Glycidyl-3-nitrobenzenesulphonate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate
OR315285-5G
APOSOR315285-5GEA
21.9
EUR
APOSOR315285-5G
(R)-Glycidyl-3-nitrobenzenesulphonate 97%
(R)-Glycidyl-3-nitrobenzenesulphonate
Formel:
C₉H₉NO₆S Molecular Weight: 259,23 g/mol Schmelzpunkt: 63 °C Flammpunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013436 CAS-Nummer: 115314-17-5 EINECS: 601-353-9 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...