(S)-(-)-ɑ-Hydroxy-γ-butyrolactone

Lieferant: Apollo Scientific

Synonyme: (S)-(-)-Dihydro-3-hydroxy-2(3H)-furanone

OR315288-1G
APOSOR315288-1GEA 944 EUR
APOSOR315288-1G
(S)-(-)-ɑ-Hydroxy-γ-butyrolactone
(S)-(-)-alpha-Hydroxy-gamma-butyrolactone
Formel: C₄H₆O₃
Molecular Weight: 102,09 g/mol
Siedepunkt: 133 °C
Dichte: 1,309 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00211245
CAS-Nummer: 52079-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR