(R)-(+)-ɑ-Hydroxy-γ-butyrolactone

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one

OR315289-1G
APOSOR315289-1GEA 270 EUR
APOSOR315289-1G
(R)-(+)-ɑ-Hydroxy-γ-butyrolactone
(R)-(+)-alpha-Hydroxy-gamma-butyrolactone
Formel: C₄H₆O₃
Molecular Weight: 102,09 g/mol
Siedepunkt: 133 °C
Dichte: 1,309 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00211246
CAS-Nummer: 56881-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR