(S,S)-(+)-2,4-Pentandiol 98%
Lieferant: Apollo Scientific
OR315298-1G
APOSOR315298-1GEA
60.9
EUR
APOSOR315298-1G
(S,S)-(+)-2,4-Pentandiol 98%
(S,S)-(+)-2,4-Pentandiol
Formel:
C₅H₁₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064276 CAS-Nummer: 72345-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...