2-Methyl-3-nitropyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR3153-1G
APOSOR3153-1GEA 26.1 EUR
APOSOR3153-1G
2-Methyl-3-nitropyridine hydrochloride
2-Methyl-3-nitropyridine hydrochloride
Formel: C₆H₇ClN₂O₂
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114123
CAS-Nummer: 63585-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR