2-Bromo-4-hydrazinylbenzonitrile 95+%

Lieferant: Apollo Scientific

OR315302-250MG
APOSOR315302-250MGEA 95.7 EUR
APOSOR315302-250MG
2-Bromo-4-hydrazinylbenzonitrile 95+%
2-Bromo-4-hydrazinylbenzonitrile
Formel: C₇H₆BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08669418
CAS-Nummer: 263845-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR