6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride 97+%

Lieferant: Apollo Scientific

OR315310-100MG
APOSOR315310-100MGEA 174 EUR
APOSOR315310-100MG
6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride 97+%
6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride
Formel: C₈H₁₁ClN₂
Molecular Weight: 170,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12913793
CAS-Nummer: 1187930-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR