3-Amino-5-methylbenzoic acid 97%

Lieferant: Apollo Scientific

OR315324-250MG
APOSOR315324-250MGEA 40.6 EUR
APOSOR315324-250MG
3-Amino-5-methylbenzoic acid 97%
3-Amino-5-methylbenzoic acid
Formel: C₈H₉NO₂
Molecular Weight: 151.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00211069
CAS-Nummer: 2305-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR