2-(2,4,5-Trichlorphenoxy)propionsäure

Lieferant: Apollo Scientific
Achtung

OR315333-25G OR315333-5G
APOSOR315333-25GEA 119 EUR
APOSOR315333-25G APOSOR315333-5G
2-(2,4,5-Trichlorphenoxy)propionsäure
2-(2,4,5-Trichlorphenoxy)propionsäure
Formel: C₉H₇Cl₃O₃
Molecular Weight: 269.51 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002646
CAS-Nummer: 93-72-1
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR