R-(-)-1-Aminoethyl phosphonic acid

Lieferant: Apollo Scientific
Achtung

OR315335-1G
APOSOR315335-1GEA 799 EUR
APOSOR315335-1G
R-(-)-1-Aminoethyl phosphonic acid
R-(-)-1-Aminoethyl phosphonic acid
Formel: C₂H₈NO₃P
Dichte: 1,48 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066502
CAS-Nummer: 60687-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR