2-(Chlormethyl)-4-methoxy-3,5-dimethylpyridinhydrochlorid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride

OR315373-5G
APOSOR315373-5GEA 60.9 EUR
APOSOR315373-5G
2-(Chlormethyl)-4-methoxy-3,5-dimethylpyridinhydrochlorid 95%
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
Formel: C₉H₁₂ClNO
Schmelzpunkt: 128 - 131 °C
MDL: MFCD00277472
CAS-Nummer: 86604-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR