3-(9H-Carbazol-9-yl)phenylboronic acid

Lieferant: Apollo Scientific

Synonyme: 3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid

OR315392-25G
APOSOR315392-25GEA 112 EUR
APOSOR315392-25G
3-(9H-Carbazol-9-yl)phenylboronic acid
3-(9H-Carbazol-9-yl)phenylboronic acid
Formel: C₁₈H₁₄BNO₂
Molecular Weight: 287.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18072506
CAS-Nummer: 864377-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR