11-Brom-1-undecanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
11-Bromoundecan-1-ol, 11-Bromoundecanol
OR315399-100G
APOSOR315399-100GEA
104
EUR
APOSOR315399-100G
11-Brom-1-undecanol
11-Brom-1-undecanol
Formel:
HO(CH₂)₁₀CH₂Br Molecular Weight: 251.21 g/mol Siedepunkt: 350 Schmelzpunkt: 46…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004752 CAS-Nummer: 1611-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...