4-Chlor-N-methylpicolinamid 97%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-N-methylpyridine-2-carboxamide
OR315405-100G
APOSOR315405-100GEA
72.5
EUR
APOSOR315405-100G
4-Chlor-N-methylpicolinamid 97%
4-Chlor-N-methylpicolinamid
Formel:
C₇H₇ClN₂O Molecular Weight: 170,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02185921 CAS-Nummer: 220000-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...