2-Boc-2,5-diazabicyclo(2.2.2)octane
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 2,5-Diazabicyclo[2.2.2]octane-2-carboxylate
OR315416-1G
APOSOR315416-1GEA
688
EUR
APOSOR315416-1G
2-Boc-2,5-diazabicyclo(2.2.2)octane
2-Boc-2,5-diazabicyclo(2.2.2)octane
Formel:
C₁₁H₂₀N₂O₂ Molecular Weight: 212,289 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18249863 CAS-Nummer: 858671-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...