2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester 95+%

Lieferant: Apollo Scientific

OR315422-100MG OR315422-1G OR315422-250MG
APOSOR315422-100MGEA 502 EUR
APOSOR315422-100MG APOSOR315422-1G APOSOR315422-250MG
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester 95+%
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester
Formel: C₂₉H₃₄N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD07779072
CAS-Nummer: 849928-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR