2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-oxo-7-azabicyclo[2.2.1]heptane-7-carboxylate

OR315428-1G
APOSOR315428-1GEA 574 EUR
APOSOR315428-1G
2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester 97%
2-Oxo-7-Azabicyclo[2.2.1]heptane-7-carboxylic acid 1,1-dimethylethyl ester
Formel: C₁₁H₁₇NO₃
Molecular Weight: 211.258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18633018
CAS-Nummer: 152533-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR