2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%

Lieferant: Apollo Scientific

OR315434-1G
APOSOR315434-1GEA 0 EUR
APOSOR315434-1G
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride
Formel: C₁₂H₂₂N₂O₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09835396
CAS-Nummer: 929302-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR