tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 1-oxo-2,7-diazaspiro[4.5]decane-7-carboxylate
OR315435-250MG
APOSOR315435-250MGEA
95.7
EUR
APOSOR315435-250MG
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate
Formel:
C₁₃H₂₂N₂O₃ Molecular Weight: 254,33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11227060 CAS-Nummer: 923009-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...