2,6-Diazaspiro[3.5]nonane-6-carboxylic acid, 1,1-dimethylethyl ester hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Diazaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester hydrochloride

OR315439-250MG
APOSOR315439-250MGEA 114 EUR
APOSOR315439-250MG
2,6-Diazaspiro[3.5]nonane-6-carboxylic acid, 1,1-dimethylethyl ester hydrochloride 97%
2,6-Diazaspiro[3.5]nonane-6-carboxylic acid, 1,1-dimethylethyl ester hydrochloride
Formel: C₁₂H₂₂N₂O₂·ClH
Molecular Weight: 262,776 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22380770
CAS-Nummer: 1279844-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR