1'-Benzylspiro[indoline-3,4'-piperidine] 95+%
Lieferant: Apollo Scientific
OR315456-250MG
APOSOR315456-250MGEA
168
EUR
APOSOR315456-250MG
1'-Benzylspiro[indoline-3,4'-piperidine] 95+%
1'-Benzylspiro[indoline-3,4'-piperidine]
Formel:
C₁₉H₂₂N₂ Molecular Weight: 278,3900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04108106 CAS-Nummer: 474538-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...