Tetraphenylphosphoniumbromid 98%

Lieferant: Apollo Scientific
Achtung

OR315491-25G OR315491-100G OR315491-500G
APOSOR315491-25GEA 50.9 EUR
APOSOR315491-25G APOSOR315491-100G APOSOR315491-500G
Tetraphenylphosphoniumbromid 98%
Tetraphenylphosphonium bromide
Formel: C₂₄H₂₀BrP
Schmelzpunkt: 294 - 296 °C
MDL: MFCD00011915
CAS-Nummer: 2751-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR