1-Brom-2,2-dimethoxypropan 97%

Lieferant: Apollo Scientific

OR315506-25G
APOSOR315506-25GEA 75.4 EUR
APOSOR315506-25G
1-Brom-2,2-dimethoxypropan 97%
1-Bromo-2,2-dimethoxypropane
Formel: BrH₂CC(OCH₃)₂CH₃
Molecular Weight: 183.05 g/mol
Siedepunkt: 83
Dichte: 1.35
Lagertemperatur: Raumtemperatur
MDL: MFCD00000208
CAS-Nummer: 126-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR