1-Boc-3-bromopiperidine 97+%

Lieferant: Apollo Scientific
Achtung

OR315514-250MG
APOSOR315514-250MGEA 0 EUR
APOSOR315514-250MG
1-Boc-3-bromopiperidine 97+%
1-Boc-3-bromopiperidine
Formel: C₁₀H₁₈BrNO₂
Siedepunkt: 296,6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07779077
CAS-Nummer: 849928-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR