1-(5-Bromo-2-furyl)ethanone

Lieferant: Apollo Scientific

OR315519-1G OR315519-250MG OR315519-500MG
APOSOR315519-1GEA 701 EUR
APOSOR315519-1G APOSOR315519-250MG APOSOR315519-500MG
1-(5-Bromo-2-furyl)ethanone
1-(5-Bromo-2-furyl)ethanone
Formel: C₆H₅BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06657513
CAS-Nummer: 3199-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR