5-Bromo-2,4-dichloropyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR315525-1G
APOSOR315525-1GEA
21.9
EUR
APOSOR315525-1G
5-Bromo-2,4-dichloropyridine
5-Bromo-2,4-dichloropyridine
Formel:
C₅H₂BrCl₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD09031035 CAS-Nummer: 849937-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...