2-Amino-4-bromobenzamide

Lieferant: Apollo Scientific
Achtung

OR315541-1G
APOSOR315541-1GEA 84.1 EUR
APOSOR315541-1G
2-Amino-4-bromobenzamide
2-Amino-4-bromobenzamide
Formel: C₇H₇BrN₂O
Molecular Weight: 215.047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674809
CAS-Nummer: 112253-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR