1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(aminomethyl)benzylcarbamate
OR315579-1G
APOSOR315579-1GEA
23.2
EUR
APOSOR315579-1G
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 95+%
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene
Formel:
C₁₃H₂₀N₂O₂ Molecular Weight: 236.3100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683058 CAS-Nummer: 108468-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...