8-Hydroxyoctanoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR315581-250MG
APOSOR315581-250MGEA
476
EUR
APOSOR315581-250MG
8-Hydroxyoctanoic acid
8-Hydroxyoctanoic acid
Formel:
C₈H₁₆O₃ Schmelzpunkt: 59…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792446 CAS-Nummer: 764-89-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...