N-Cyclopropyl-N-(4-piperidinyl)benzenesulphonamide 95+%

Lieferant: Apollo Scientific
Achtung

OR3156-1G
APOSOR3156-1GEA 112 EUR
APOSOR3156-1G
N-Cyclopropyl-N-(4-piperidinyl)benzenesulphonamide 95+%
N-Cyclopropyl-N-(4-piperidinyl)benzenesulphonamide
Formel: C₁₄H₂₀N₂O₂S
Molecular Weight: 280.39 g/mol
Schmelzpunkt: >150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567982
CAS-Nummer: 387350-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR