1,4-Bis(diphenylphosphino)butan 97%

Lieferant: Apollo Scientific

Synonyme: Butan-1,3-diylbis(diphenylphosphin), Butylenbis(diphenylphosphin), Bis(1,2-diphenylphosphino)butan

OR315705-100G
APOSOR315705-100GEA 34.8 EUR
APOSOR315705-100G
1,4-Bis(diphenylphosphino)butan 97%
1,4-Bis(diphenylphosphino)butan
Formel: C₂₈H₂₈P₂
Molecular Weight: 426,48 g/mol
Schmelzpunkt: 132…133,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003051
CAS-Nummer: 7688-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR