3-(Bromoacetyl)-5-chloro-2-thiophenesulphonamide 95+%

Lieferant: Apollo Scientific

Synonyme: 3-(2-Bromoacetyl)-5-chlorothiophene-2-sulphonamide

OR315747-5G
APOSOR315747-5GEA 87 EUR
APOSOR315747-5G
3-(Bromoacetyl)-5-chloro-2-thiophenesulphonamide 95+%
3-(Bromoacetyl)-5-chloro-2-thiophenesulphonamide
Formel: C₆H₅BrClNO₃S₂
Molecular Weight: 318,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09033310
CAS-Nummer: 160982-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR