4,4'-Bis(chlormethyl)biphenyl
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4,4-Bis(chloromethyl)-1,1-biphenyl, 4,4'-Bis(chloromethyl)-1,1-biphenyl
OR315763-500G
APOSOR315763-500GEA
98.6
EUR
APOSOR315763-500G
4,4'-Bis(chlormethyl)biphenyl
4,4'-Bis(chlormethyl)biphenyl
Formel:
C₁₄H₁₂Cl₂ Molecular Weight: 251.15 g/mol Schmelzpunkt: 126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674019 CAS-Nummer: 1667-10-3 EINECS: 216-784-4 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...