3-Amino-2-naphthoesäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-naphthalincarbonsäure, 3-Aminonaphthalin-2-carbonsäure

OR315769-5G OR315769-25G
APOSOR315769-5GEA 60.9 EUR
APOSOR315769-5G APOSOR315769-25G
3-Amino-2-naphthoesäure 97%
3-Amino-2-naphthoesäure
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004115
CAS-Nummer: 5959-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR