Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl) 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl
OR315773-100G
OR315773-25G
OR315773-5G
APOSOR315773-100GEA
157
EUR
APOSOR315773-100G
APOSOR315773-25G
APOSOR315773-5G
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl) 98%
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Formel:
C₆H₁₄ClNO₂ Schmelzpunkt: 141…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058255 CAS-Nummer: 27532-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...