trans-4-Aminoadamantan-1-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315777-1G
APOSOR315777-1GEA
33.5
EUR
APOSOR315777-1G
trans-4-Aminoadamantan-1-ol 95%
trans-4-Aminoadamantan-1-ol
Formel:
C₁₀H₁₇NO Molecular Weight: 167,248 g/mol Schmelzpunkt: 230…235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12761603 CAS-Nummer: 62058-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...