(1S,3S,4S)-4-Amino-1-adamantanolhydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
OR315778-5G
APOSOR315778-5GEA
84.1
EUR
APOSOR315778-5G
(1S,3S,4S)-4-Amino-1-adamantanolhydrochlorid
trans-4-Aminoadamantan-1-ol hydrochloride
Formel:
C₁₀H₁₈ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11099880 CAS-Nummer: 62075-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...