Adamantan-1-yl acrylate

Lieferant: Apollo Scientific

OR315783-1G OR315783-5G
APOSOR315783-1GEA 0 EUR
APOSOR315783-1G APOSOR315783-5G
Adamantan-1-yl acrylate
Adamantan-1-yl acrylate
Formel: C₁₃H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18072700
CAS-Nummer: 121601-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR