3-Bromothiophene-4-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR315853-250MG OR315853-1G
APOSOR315853-250MGEA 219 EUR
APOSOR315853-250MG APOSOR315853-1G
3-Bromothiophene-4-sulphonyl chloride 95%
3-Bromothiophene-4-sulphonyl chloride
Formel: C₄H₂BrClO₂S₂
Molecular Weight: 261.54 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04115406
CAS-Nummer: 111283-90-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR