2-Bromo-3,5-dichlorophenol 95%

Lieferant: Apollo Scientific

OR315876-1G
APOSOR315876-1GEA 857 EUR
APOSOR315876-1G
2-Bromo-3,5-dichlorophenol 95%
2-Bromo-3,5-dichlorophenol
Formel: C₆H₃BrCl₂O
Molecular Weight: 241,897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22414726
CAS-Nummer: 13659-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR