3-Bromo-4-chloro-2-hydroxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR315877-1G OR315877-500MG
APOSOR315877-1GEA 1060 EUR
APOSOR315877-1G APOSOR315877-500MG
3-Bromo-4-chloro-2-hydroxybenzaldehyde 95%
3-Bromo-4-chloro-2-hydroxybenzaldehyde
Formel: C₇H₄BrClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23705580
CAS-Nummer: 872274-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR