1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride 95%

Lieferant: Apollo Scientific

OR315878-25MG
APOSOR315878-25MGEA 509 EUR
APOSOR315878-25MG
1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride 95%
1-(3-(Piperazin-1-yl)azetidin-1-yl)ethanone hydrochloride
Formel: C₉H₁₇N₃O·ClH
Molecular Weight: 219,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30489690
CAS-Nummer: 2208138-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR