5-Amino-4,6-dichloronicotinonitrile 95%
Lieferant: Apollo Scientific
OR315879-25MG
APOSOR315879-25MGEA
857
EUR
APOSOR315879-25MG
5-Amino-4,6-dichloronicotinonitrile 95%
5-Amino-4,6-dichloronicotinonitrile
Formel:
C₆H₃Cl₂N₃ Molecular Weight: 188,014 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30293239 CAS-Nummer: 1565871-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...