Methyl 4-(cyclopropyl)thiophene-2-carboxylate 95%

Lieferant: Apollo Scientific

OR315881-1G OR315881-250MG
APOSOR315881-1GEA 574 EUR
APOSOR315881-1G APOSOR315881-250MG
Methyl 4-(cyclopropyl)thiophene-2-carboxylate 95%
Methyl 4-(cyclopropyl)thiophene-2-carboxylate
Formel: C₉H₁₀O₂S
Molecular Weight: 182.24 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32689351
CAS-Nummer: 2111296-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR