3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%

Lieferant: Apollo Scientific

OR315883-1G OR315883-250MG OR315883-5G
APOSOR315883-1GEA 574 EUR
APOSOR315883-1G APOSOR315883-250MG APOSOR315883-5G
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
3-(Cyclopropyl)thiophene-2-carboxaldehyde
Formel: C₈H₈OS
Lagertemperatur: Kühlschrank
MDL: MFCD22070104
CAS-Nummer: 29481-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR