3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
Lieferant: Apollo Scientific
OR315883-1G
OR315883-250MG
OR315883-5G
APOSOR315883-1GEA
574
EUR
APOSOR315883-1G
APOSOR315883-250MG
APOSOR315883-5G
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
3-(Cyclopropyl)thiophene-2-carboxaldehyde
Formel:
C₈H₈OS Lagertemperatur: Kühlschrank |
MDL:
MFCD22070104 CAS-Nummer: 29481-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...